Merck
  • The MP65 gene is required for cell wall integrity, adherence to epithelial cells and biofilm formation in Candida albicans.
The MP65 gene is required for cell wall integrity, adherence to epithelial cells and biofilm formation in Candida albicans.
BMC microbiology (2011-05-18)
Silvia Sandini, Annarita Stringaro, Silvia Arancia, Marisa Colone, Francesca Mondello, Susanna Murtas, Antonietta Girolamo, Nicolina Mastrangelo, Flavia De Bernardis
ABSTRACT
The MP65 gene of Candida albicans (orf19.1779) encodes a putative β-glucanase mannoprotein of 65 kDa, which plays a main role in a host-fungus relationship, morphogenesis and pathogenicity. In this study, we performed an extensive analysis of a mp65Δ mutant to assess the role of this protein in cell wall integrity, adherence to epithelial cells and biofilm formation. The mp65Δ mutant showed a high sensitivity to a range of cell wall-perturbing and degrading agents, especially Congo red, which induced morphological changes such as swelling, clumping and formation of hyphae. The mp65Δ mutant showed an activation of two MAPKs (Mkc1p and Cek1p), a high level of expression of two stress-related genes (DDR48 and SOD5), and a modulated expression of β-glucan epitopes, but no gross changes in cell wall polysaccharide composition. Interestingly, the mp65Δ mutant displayed a marked reduction in adhesion to BEC and Caco-2 cells and severe defects in biofilm formation when compared to the wild type. All of the mentioned properties were totally or partially recovered in a revertant strain, demonstrating the specificity of gene deletion. We demonstrate that the MP65 gene of Candida albicans plays a significant role in maintaining cell wall integrity, as well as in adherence to epithelia and biofilm formation, which are major virulence attributes of this fungus.
Sigma-Aldrich
Phosphate buffered saline, pH 7.2 (25 °C)
Sigma-Aldrich
Phosphate buffered saline, Autoclaved, pH 7.2 (25 °C)
Sigma-Aldrich
Phosphate buffered saline, pH 7.4, contains BSA, powder
Supelco
Congo Red, analytical standard
Sigma-Aldrich
Phosphate buffered saline, BioUltra, solution
Sigma-Aldrich
Phosphate buffered saline, BioUltra, pH 7.4 ( in solution), contains TWEEN 20, tablet
Sigma-Aldrich
Phosphate buffered saline, tablet
Sigma-Aldrich
Phosphate buffered saline, BioUltra, washing buffer for peroxidase conjugates in Western Blotting, 10x concentrate
Sigma-Aldrich
Phosphate Buffered Saline, 10× PBS for Western blots and IP
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Phosphate buffered saline, powder, pH 7.4, for preparing 1 L solutions
Sigma-Aldrich
Phosphate buffered saline, 10× concentrate, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
Phosphate buffered saline, pH 7.6 (25 °C)
Sigma-Aldrich
Phosphate Buffered Saline with 3% Non-Fat Milk, pH 7.4
Sigma-Aldrich
Phosphate buffered saline, with 5% nonfat milk, powder (dry milled), pH 7.3
Sigma-Aldrich
Phosphate buffered saline, powder, pH 7.4, for preparing 5 L solutions
Sigma-Aldrich
Phosphate Buffered Saline with 0.05% TWEEN® 20, pH 7.4
Sigma-Aldrich
Phosphate buffered saline, BioPerformance Certified, pH 7.4
Sigma-Aldrich
Congo Red, certified by the BSC, BioXtra
Sigma-Aldrich
Congo Red, Dye content ≥35 %
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Calcofluor White Stain, suitable for microbiology