Merck
  • A pseudoautosomal glycosylation disorder prompts the revision of dolichol biosynthesis.
A pseudoautosomal glycosylation disorder prompts the revision of dolichol biosynthesis.
Cell (2024-06-01)
Matthew P Wilson, Takfarinas Kentache, Charlotte R Althoff, Céline Schulz, Geoffroy de Bettignies, Gisèle Mateu Cabrera, Loreta Cimbalistiene, Birute Burnyte, Grace Yoon, Gregory Costain, Sandrine Vuillaumier-Barrot, David Cheillan, Daisy Rymen, Lucie Rychtarova, Hana Hansikova, Marina Bury, Joseph P Dewulf, Francesco Caligiore, Jaak Jaeken, Vincent Cantagrel, Emile Van Schaftingen, Gert Matthijs, François Foulquier, Guido T Bommer
ABSTRACT
Dolichol is a lipid critical for N-glycosylation as a carrier for activated sugars and nascent oligosaccharides. It is commonly thought to be directly produced from polyprenol by the enzyme SRD5A3. Instead, we found that dolichol synthesis requires a three-step detour involving additional metabolites, where SRD5A3 catalyzes only the second reaction. The first and third steps are performed by DHRSX, whose gene resides on the pseudoautosomal regions of the X and Y chromosomes. Accordingly, we report a pseudoautosomal-recessive disease presenting as a congenital disorder of glycosylation in patients with missense variants in DHRSX (DHRSX-CDG). Of note, DHRSX has a unique dual substrate and cofactor specificity, allowing it to act as a NAD+-dependent dehydrogenase and as a NADPH-dependent reductase in two non-consecutive steps. Thus, our work reveals unexpected complexity in the terminal steps of dolichol biosynthesis. Furthermore, we provide insights into the mechanism by which dolichol metabolism defects contribute to disease.
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
ANTI-DHRSX antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution