Merck
  • Inhibition of HMG CoA reductase reveals an unexpected role for cholesterol during PGC migration in the mouse.
Inhibition of HMG CoA reductase reveals an unexpected role for cholesterol during PGC migration in the mouse.
BMC developmental biology (2009-01-02)
Jiaxi Ding, Dechen Jiang, Michael Kurczy, Jennifer Nalepka, Brian Dudley, Erin I Merkel, Forbes D Porter, Andrew G Ewing, Nicholas Winograd, James Burgess, Kathleen Molyneaux
ABSTRACT
Primordial germ cells (PGCs) are the embryonic precursors of the sperm and eggs. Environmental or genetic defects that alter PGC development can impair fertility or cause formation of germ cell tumors. We demonstrate a novel role for cholesterol during germ cell migration in mice. Cholesterol was measured in living tissue dissected from mouse embryos and was found to accumulate within the developing gonads as germ cells migrate to colonize these structures. Cholesterol synthesis was blocked in culture by inhibiting the activity of HMG CoA reductase (HMGCR) resulting in germ cell survival and migration defects. These defects were rescued by co-addition of isoprenoids and cholesterol, but neither compound alone was sufficient. In contrast, loss of the last or penultimate enzyme in cholesterol biosynthesis did not alter PGC numbers or position in vivo. However embryos that lack these enzymes do not exhibit cholesterol defects at the stage at which PGCs are migrating. This demonstrates that during gestation, the cholesterol required for PGC migration can be supplied maternally. In the mouse, cholesterol is required for PGC survival and motility. It may act cell-autonomously by regulating clustering of growth factor receptors within PGCs or non cell-autonomously by controlling release of growth factors required for PGC guidance and survival.
Sigma-Aldrich
Mevinolin from Aspergillus sp., ≥98% (HPLC)
Sigma-Aldrich
11-Mercaptoundecanoic acid, 95%
Sigma-Aldrich
11-Mercaptoundecanoic acid, 98%
Sigma-Aldrich
Farnesol, mixture of isomers, ≥95%, stabilized, FG
Sigma-Aldrich
Farnesol, 95%
Sigma-Aldrich
Geranylgeraniol, ≥85% (GC)
Sigma-Aldrich
Polyacrylamide, nonionic water-soluble polymer
Sigma-Aldrich
Polyacrylamide, average Mn 150,000