Merck
  • Ratiometrically pH-Insensitive Upconversion Nanoprobe: Toward Simultaneously Quantifying Organellar Calcium and Chloride and Understanding the Interaction of the Two Ions in Lysosome Function.
Ratiometrically pH-Insensitive Upconversion Nanoprobe: Toward Simultaneously Quantifying Organellar Calcium and Chloride and Understanding the Interaction of the Two Ions in Lysosome Function.
Analytical chemistry (2022-07-26)
Caiping Ding, Shasha Cheng, Fang Yuan, Cuiling Zhang, Yuezhong Xian
ABSTRACT
Calcium and chloride levels are closely related to lysosome dysfunction. However, the simultaneous measurement of calcium (Ca2+) and chloride (Cl-) in acidic subcellular organelles, which is conducive to a deep understanding of lysosome-related biological events, remains a challenge. In this study, we developed a pH-insensitive, ratiometric NIR nanoprobe for the simultaneous detection of Ca2+ and Cl- in acidic lysosomes and determined the roles of the two ions in lysosome function. The upconversion nanoprobe with blue, green, and red emissions was modified with a Ca2+-sensitive dye (Rhod-5N) and Cl--responsive fluorophore (10,10'-bis[3-carboxypropyl]-9,9'-biacridinium dinitrate, BAC). As a result of a dual-luminescence resonance energy transfer between upconversion nanoparticles (UCNPs) and Rhod-5N/BAC, the blue and green upconversion luminescence (UCL) of UCNPs were quenched and the red UCL was used as the reference signal. The ratiometric upconversion nanoprobe possesses a specific ability for the concurrent recognition of Ca2+ and Cl- ions independent of the influence of the environmental pH. To locate the probe in the lysosome, dextran was further modified with upconversion nanoparticles. Then, the nanoprobe with a high spatial resolution was constructed for the simultaneous monitoring of Ca2+ and Cl- in acidic lysosomes. Moreover, it was found that the reduction of lysosomal Cl- affects the release of lysosomal Ca2+, which further blocks the activities of specific lysosomal enzymes. The ratiometric NIR nanoprobe has great potential for decoding and evaluating lysosomal diseases.
Product Description
Rhod-5N potassium salt, ≥95% (HPLC)