Merck
  • Mortality in human sepsis is associated with downregulation of Toll-like receptor 2 and CD14 expression on blood monocytes.
Mortality in human sepsis is associated with downregulation of Toll-like receptor 2 and CD14 expression on blood monocytes.
Diagnostic pathology (2009-04-18)
Bernhard Schaaf, Karen Luitjens, Torsten Goldmann, Tobias van Bremen, Friedhelm Sayk, Christoph Dodt, Klaus Dalhoff, Daniel Droemann
ABSTRACT
Pattern recognition receptors are a key component of the first line host defense against infection, recognizing specific microbial products. We hypothesize that monocyte hyporesponsiveness in human sepsis is associated with a downregulation of the pattern recognition receptors Toll-like receptor (TLR)-2 and TLR4.Protein expression of CD14, TLR2 and TLR4 on blood monocytes was examined using flow cytometry from 29 patients with sepsis and 14 healthy controls. In addition LPS stimulated TNF-alpha and IL-10 production was studied in a 24 hour whole blood assay.We found an increased expression of CD14, TLR2 and TLR4 in patients with sepsis compared to controls (p < 0.01). In patients with sepsis, death was associated with significant lower CD14 and TLR2 expression at admission (CD14: 25.7 +- 19.1 vs 39.1 +- 17.3 mean fluorescence intensity [MFI], p = 0.02; TLR2: 21.8 +- 9.4 vs. 30.9 +- 9.6, p = 0.01). At 72 hours the TLR2 expression on monocytes was associated with the IL-10 inducibility after LPS stimulation (r = 0.52, p = 0.02) and the CD14 expression with the IL-6, IL-10 and TNF inducibility.We conclude that septic patients are characterized by an increased expression of CD14, TLR2 and TLR4 on monocytes compared to controls. Death is associated with downregulation of TLR2 and CD14 expression on monocytes correlating with reduced cytokine inducibility. We suggest that CD14 and TLR2 are a key factor in monocyte hyporesponsibility during severe sepsis.
Sigma-Aldrich
Lipoteichoic acid from Staphylococcus aureus, bacterial cell wall polymer
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O26:B6, purified by gel-filtration chromatography
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O26:B6, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O26:B6, ≥10,000 EU/mg, purified by phenol extraction
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O26:B6, purified by trichloroacetic acid extraction
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O26:B6, Ready Made solution, 1 mg/mL, 0.2 μm filtered