Merck
The diamidine DB75 targets the nucleus of Plasmodium falciparum.
Malaria journal (2009-05-16)
Anne E Purfield, Richard R Tidwell, Steven R Meshnick
ABSTRACT
DB289, [2,5-bis(4-amidinophenyl)furan bis-O-methylamidoxime], is a broad spectrum anti-parasitic compound which has been shown to be effective against malaria in recent clinical trials. DB75, [2,5-bis(4-amidinophenyl)furan], is the active metabolite of this drug. The objective of this study was to determine the mechanism of action of DB75 in Plasmodium falciparum. Live parasites were observed by confocal microscopy after treatment with organelle specific dyes and DB75, an inherently fluorescent compound. Parasites were exposed to DB75 and assessed for growth and morphological changes over time using blood smears and light microscopy. Also, to determine if DB75 affects gene transcription, real time PCR was used to monitor transcript levels over time for six developmentally expressed genes, including trophozoite antigen R45-like (PFD1175w), lactate dehydrogenase (PF13_0141), DNA primase (PFI0530c), isocitrate dehydrogenase (PF13_0242), merozoite surface protein-1 (PFI1475w), and merozoite surface protein-7 (PF13_0197). The results show that DB75 localizes in the parasite nucleus but not in other organelles. Once rings are exposed, parasites mature to the trophozoite stage and stall. No stage-dependent or gene-specific inhibition of transcription was seen. However, DB75 delayed peak transcription of trophozoite-stage genes. Taken together, DB75 appears to concentrate in the nucleus and delay parasite maturation.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material