Merck
  • Role of glycocalyx in flow-induced production of nitric oxide and reactive oxygen species.
Role of glycocalyx in flow-induced production of nitric oxide and reactive oxygen species.
Free radical biology & medicine (2009-06-09)
Robert Kumagai, Xiao Lu, Ghassan S Kassab
ABSTRACT
Although the glycocalyx has been implicated in wall shear stress (WSS) mechanotransduction, the role of glycocalyx components in nitric oxide (NO) and reactive oxygen species (ROS) production remains unclear. Here, we tested the hypothesis that glycocalyx is implicated in both endothelial NO and O(2)(-) production. Specifically, we evaluated the role of hyaluronic acid (HA), heparan sulfate (HS), and sialic acid (SA) in NO and O(2)(-) mechanotransduction. Twenty-seven ex vivo porcine superficial femoral arteries were incubated with heparinase III, hyaluronidase, or neuraminidase, to remove HS, HA, or SA, respectively, from glycocalyx. The arteries were then subjected to steady-state flow and the effluent solution was measured for nitrites and the vessel diameter was tracked to quantify the degree of vasodilation. Our results show that removal of HA decreased both nitrites and vasodilation, and tempol treatment had no reversing effect. Degradation of HS proteoglycans decreased NO bioavailability through an increase in O(2)(-) production as indicated by fluorescent signals of dihydroethidium (DHE) and its area fraction (209+/-24% increase) and also removed extracellular O(2)(-) dismutase (ecSOD) (67+/-9% decrease). The removal of SA also increased O(2)(-) production as indicated by DHE fluorescent signals (86+/-17% increase) and the addition of tempol, a mimic O(2)(-) scavenger, restored both NO availability and vasodilation in both heparinase- and neuraminidase-treated vessels. This implies that HS and SA are not directly involved in WSS-mediated NO production. This study implicates HA in WSS-mediated NO mechanotransduction and underscores the role of HS and SA in ROS regulation in vessel walls in response to WSS stimulation.
Sigma-Aldrich
Albumin from porcine serum, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from porcine serum, lyophilized powder, 96-99% (agarose gel electrophoresis)
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, lyophilized powder (essentially salt-free), 750-3000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid
Sigma-Aldrich
Albumin from porcine serum, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, pH <5.0, powder
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free and essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, powder, BioXtra
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, 1× crystallized, ≥97% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, lyophilized powder, essentially fatty acid free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, suitable for hybridization, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, suitable for RIA, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, low endotoxin, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, cold ethanol fraction, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, low endotoxin, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, microbiologically tested, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥97% (agarose gel electrophoresis), 1X Crystallized
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, BioReagent, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, dimer mol wt ~132 kDa
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, suitable for (for molecular biology), Non-acetylated, Molecular Biology