Merck
  • An improved protocol for quantification of freshwater Actinobacteria by fluorescence in situ hybridization.
An improved protocol for quantification of freshwater Actinobacteria by fluorescence in situ hybridization.
Applied and environmental microbiology (2003-05-07)
Raju Sekar, Annelie Pernthaler, Jakob Pernthaler, Falk Warnecke, Thomas Posch, Rudolf Amann
ABSTRACT
We tested a previously described protocol for fluorescence in situ hybridization of marine bacterioplankton with horseradish peroxidase-labeled rRNA-targeted oligonucleotide probes and catalyzed reporter deposition (CARD-FISH) in plankton samples from different lakes. The fraction of Bacteria detected by CARD-FISH was significantly lower than after FISH with fluorescently monolabeled probes. In particular, the abundances of aquatic Actinobacteria were significantly underestimated. We thus developed a combined fixation and permeabilization protocol for CARD-FISH of freshwater samples. Enzymatic pretreatment of fixed cells was optimized for the controlled digestion of gram-positive cell walls without causing overall cell loss. Incubations with high concentrations of lysozyme (10 mg ml(-1)) followed by achromopeptidase (60 U ml(-1)) successfully permeabilized cell walls of Actinobacteria for subsequent CARD-FISH both in enrichment cultures and environmental samples. Between 72 and >99% (mean, 86%) of all Bacteria could be visualized with the improved assay in surface waters of four lakes. For freshwater samples, our method is thus superior to the CARD-FISH protocol for marine Bacteria (mean, 55%) and to FISH with directly fluorochrome labeled probes (mean, 67%). Actinobacterial abundances in the studied systems, as detected by the optimized protocol, ranged from 32 to >55% (mean, 45%). Our findings confirm that members of this lineage are among the numerically most important Bacteria of freshwater picoplankton.
Sigma-Aldrich
Achromopeptidase from bacteria, partially purified powder, ≥20,000 units/mg solid
Sigma-Aldrich
Akkermansia muciniphila FISH probe-ATTO488, Probe for fluorescence in situ hybridization (FISH), 20µM in water
Sigma-Aldrich
Escherichia coli FISH probe -ATTO488, Probe for fluorescence in situ hybridization (FISH), 20µM in water
Sigma-Aldrich
Enterococcus faecium FISH probe - Cy3, Probe for fluorescence in situ hybridization (FISH), 20µM in water
Sigma-Aldrich
Enterococcus faecium FISH probe – ATTO 488, Probe for fluorescence in situ hybridization (FISH), 20µM in water
Sigma-Aldrich
Eubacteria FISH probe - ATTO488, Probe for fluorescence in situ hybridization (FISH), 20 µM in water
Sigma-Aldrich
Porphyromonas gingivalis FISH probe - Cy3, Probe for fluorescence in situ hybridization (FISH),20 μM in water
Sigma-Aldrich
Bacterial negative control FISH probe - Cy3, Probe for fluorescence in situ hybridization (FISH),20 μM in water
Sigma-Aldrich
Proteus FISH probe - Cy3, Probe for fluorescence in situ hybridization (FISH),20 μM in water
Sigma-Aldrich
Alphaproteobacteria FISH probe-Cy3, Probe for fluorescence in situ hybridization (FISH), 20 μM in water
Sigma-Aldrich
Eubacteria FISH probe - Cy3, Probe for fluorescence in situ hybridization (FISH)
Sigma-Aldrich
Escherichia coli FISH probe - Cy3, Probe for fluorescence in situ hybridization (FISH)
Sigma-Aldrich
Porphyromonas gingivalis FISH probe - ATTO488, Probe for fluorescence in situ hybridization (FISH)
Sigma-Aldrich
Bacterial negative control FISH probe - ATTO488, Probe for fluorescence in situ hybridization (FISH),20 μM in water
Sigma-Aldrich
Proteus FISH probe - ATTO488, Probe for fluorescence in situ hybridization (FISH),20 μM in water
Sigma-Aldrich
Akkermansia muciniphila FISH probe - Cy3, Probe for fluorescence in situ hybridization (FISH),20 μM in water