Skip to Content
AT
EN
Products
Applications
Services
Resources
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Lab Automation
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Documents
Learning Center
New
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Home
Search Results
Isoelectric focusing.
Isoelectric focusing.
Methods in enzymology (1990-01-01)
D E Garfin
PMID
2314254
Product Description
S3889
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Pricing
S6021
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), Molecular Biology
Pricing
S7547
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Pricing
S1876
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Pricing
85529
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Pricing
240850
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Pricing