Merck
  • Inhibition of Staphylococcus aureus by lysostaphin-expressing Lactobacillus plantarum WCFS1 in a modified genital tract secretion medium.
Inhibition of Staphylococcus aureus by lysostaphin-expressing Lactobacillus plantarum WCFS1 in a modified genital tract secretion medium.
Applied and environmental microbiology (2011-10-11)
Huanli Liu, Yuan Gao, Li-Rong Yu, Richard C Jones, Christopher A Elkins, Mark E Hart
ABSTRACT
Lactobacillus species are a predominant member of the vaginal microflora and are critical in maintaining an acidic vaginal environment thought to contribute to the prevention of a number of urogenital diseases. However, during menstruation the pH of the vaginal environment increases to neutrality, a pH conducive for Staphylococcus aureus proliferation and the production of toxic shock syndrome toxin 1 (TSST-1) in susceptible women. In order to generate Lactobacillus species capable of expressing lysostaphin (an endopeptidase that cleaves the cell wall of S. aureus) in a modified genital tract secretion medium (mGTS) under neutral-pH conditions, six prominent proteins from Lactobacillus plantarum WCFS1 spent medium were identified by mass spectrometry. Sequences for promoters, signal peptides, and mature lysostaphin were used to construct plasmids that were subsequently transformed into L. plantarum WCFS1. The promoter and signal sequences of Lp_3014 (putatively identified as a transglycosylase) or the promoter sequence of Lp_0789 (putatively identified as glyceraldehyde 3-phosphate dehydrogenase) with the signal sequence of Lp_3014 exhibited lysostaphin activity on buffered medium containing heat-killed S. aureus. The cassettes were integrated into the chromosome of L. plantarum WCFS1, but only the cassette containing the promoter and signal sequence from Lp_3014 had integrated into the appropriate site. Coculture assays using buffered mGTS showed that lysostaphin expressed from L. plantarum WCFS1 reduced the growth of TSST-1-producing strains of S. aureus under neutral-pH conditions. This study provides the basis for determining whether lysostaphin-producing Lactobacillus strains could potentially be used as a means to inhibit the growth of S. aureus during menstruation.
Sigma-Aldrich
Lysostaphin from Staphylococcus simulans, recombinant, expressed in E. coli, lyophilized powder
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, BioUltra, ≥97% (SDS-PAGE), Protein 40-60 % by biuret, ≥2,000 units/mg protein
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, lyophilized powder, Protein 50-70 % by biuret, ≥500 units/mg protein
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, aseptically filled