Merck
  • Specific actions of GLP-1 receptor agonists and DPP4 inhibitors for the treatment of pancreatic β-cell impairments in type 2 diabetes.
Specific actions of GLP-1 receptor agonists and DPP4 inhibitors for the treatment of pancreatic β-cell impairments in type 2 diabetes.
Cellular signalling (2012-11-20)
Stéphane Dalle, Rémy Burcelin, Pierre Gourdy
ABSTRACT
Type 2 diabetes occurs when the β-cells do not secrete enough insulin to counter balance insulin resistance. GLP-1 and GIP are insulinotropic peptides which are thought to benefit to β-cell physiology. On one hand sustained pharmacological levels of GLP-1 are achieved by subcutaneous administration of GLP-1 analogs while transient and lower physiological levels of GLP-1 are attained following DPP4 inhibitor (DPP4i) treatment. On the other hand, DPP4i increase GLP-1 concentration into the portal vein to recruit the gut-to brain-to pancreas axis which is not the case with injected analogs. Hence, these differences between GLP-1 analogs and DPP4i indicate that both strategies could differentially impact β-cell behavior. Here, we summarize the effects of GLP-1 analogs and DPP4i on β-cell physiology. We discuss the possibility that production of signaling molecules, such as cAMP, generated into the β-cells by native GLP-1 or pharmacological GLP-1 analogs may vary and engage different downstream signaling networks. Hence, deciphering which signaling networks are engaged following GLP-1 analogs or DPP4i administration appears to be critical to unveil the contribution of each treatment/strategy to engage β-cell cellular processes.
Sigma-Aldrich
Dipeptidyl Peptidase IV human, recombinant, expressed in Sf9 cells
Sigma-Aldrich
Dipeptidyl Peptidase IV human, recombinant, expressed in baculovirus infected Sf9 cells, pkg of ≥1.0 units/vial, ≥10 units/mg protein