Merck
  • Combination of liquid hot water pretreatment and wet disk milling to improve the efficiency of the enzymatic hydrolysis of eucalyptus.
Combination of liquid hot water pretreatment and wet disk milling to improve the efficiency of the enzymatic hydrolysis of eucalyptus.
Bioresource technology (2012-12-25)
Wei Weiqi, Wu Shubin, Liu Liguo
ABSTRACT
Combination of liquid hot water pretreatment (LHWP) and wet disk milling (WDM) was investigated in this study to enhance the sugar recovery yield both in prehydrolyzate and enzymatic hydrolyzate. The results show that WDM with LHWP at 180 °C for 20 min produced maximum xylose and glucose yields of 91.62% and 88.12%, respectively, which are higher than that of dilute acid pretreatment or individual LHWP. Corresponding concentration of fermentation inhibitors such as acetic acid, HMF, and furfural in the prehydrolyzate are about 0.98, 0.07 and 0.78 g/L, respectively, which indicated that the detoxification may be not required in the next fermentation step. The acid-insoluble lignin recovery in the insoluble solid resulting from enzymatic hydrolysis, was 25.67/100g raw material, representing 90.7% of acid-insoluble lignin in the eucalyptus biomass. It can be concluded that liquid hot water pretreatment combined with wet disk milling can be successfully applied to eucalyptus.
Sigma-Aldrich
Cellulase from Trichoderma reesei, aqueous solution, ≥700 units/g
Sigma-Aldrich
Cellulase from Trichoderma reesei ATCC 26921, lyophilized powder, ≥1 unit/mg solid
Sigma-Aldrich
Cellulase from Trichoderma sp., BioReagent, suitable for plant cell culture, 3-10 units/mg solid
Sigma-Aldrich
Cellulase from Trichoderma sp., powder, ≥5,000 units/g solid
Sigma-Aldrich
Cellulase from Aspergillus niger, powder, off-white, ~0.8 U/mg
Sigma-Aldrich
Cellulase from Aspergillus niger, powder, ≥0.3 units/mg solid
Sigma-Aldrich
Cellulase from Aspergillus sp., aqueous solution