Merck
PDK1 inhibition is a novel therapeutic target in multiple myeloma.
British journal of cancer (2013-01-17)
S Fujiwara, Y Kawano, H Yuki, Y Okuno, K Nosaka, H Mitsuya, H Hata
ABSTRACT
Cancer cells utilise the glycolytic pathway even when adequate oxygen is present, a phenomenon known as the Warburg effect. We examined whether this system is operative in multiple myeloma (MM) cells and whether glycolysis inhibition is a potential therapeutic modality. The MM cells were purified from 59 patients using CD138-immunomagnetic beads. The expression levels of genes associated with glycolysis, c-MYC, GLUT1, LDHA, HIF1A and pyruvate dehydrogenase kinase-1 (PDK1) were determined by real-time PCR. Glucose consumption and lactate production by MM cell lines were analysed. Oxamate, an LDH inhibitor, and dichloroacetate (DCA), a PDK1 inhibitor, were employed. Inhibition of PDK1 expression was achieved using a siRNA. High LDHA expression was found to be an indicator of poor prognosis. It was also positively correlated with the expression of PDK1, c-MYC and GLUT1. Greater glucose consumption and lactate production in MM cells was associated with higher LDHA expression. All the glycolysis inhibitors (oxamate, DCA and PDK1 siRNA) induced apoptosis in MM cells. DCA combined with bortezomib showed additive cytotoxic effects. The present data suggest that the Warburg effect is operative in MM cells. As PDK1 is not overexpressed in normal tissues, PDK1 inhibition could serve as a novel therapeutic approach.
Sigma-Aldrich
D-Lactic Dehydrogenase from Staphylococcus epidermidis, lyophilized powder, ≥80 units/mg solid
Sigma-Aldrich
Sodium oxamate, ≥98%
Sigma-Aldrich
Dichloroacetic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
D-Lactic Dehydrogenase from Lactobacillus leichmannii, lyophilized powder, 150-500 units/mg protein
Sigma-Aldrich
D-Lactic Dehydrogenase from Lactobacillus leichmannii, ammonium sulfate suspension, ≥250 units/mg protein (biuret)
Sigma-Aldrich
DCA Deblock (0.36M dichloroacetic acid in toluene)
Supelco
Dichloroacetic acid, PESTANAL®, analytical standard