Merck
  • Locally-generated acetaldehyde is involved in ethanol-mediated LTP inhibition in the hippocampus.
Locally-generated acetaldehyde is involved in ethanol-mediated LTP inhibition in the hippocampus.
Neuroscience letters (2013-01-29)
Kazuhiro Tokuda, Yukitoshi Izumi, Charles F Zorumski
ABSTRACT
Consistent with the ability of severe alcohol intoxication to impair memory, high concentrations of ethanol (60mM) acutely inhibit long-term potentiation (LTP) in the CA1 region of rat hippocampal slices. To account for this, we hypothesized that local metabolism to acetaldehyde may contribute to the effects of high ethanol on synaptic function. However, sodium azide, a catalase inhibitor, and allyl sulfide, an inhibitor of cytochrome P450 2E1 (CYP2E1), failed to overcome LTP inhibition by 60mM ethanol. In contrast, LTP was successfully induced in the presence of ethanol plus 4-methylpyrazole (4MP), an inhibitor of alcohol dehydrogenase, suggesting that local metabolism via alcohol dehydrogenase contributes to synaptic effects. Furthermore, exogenously administered acetaldehyde overcame the effects of 4MP on LTP inhibition mediated by ethanol. These observations indicate that acetaldehyde generated by local metabolism within the hippocampus participates in the synaptic dysfunction associated with severe alcohol intoxication.
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Allyl sulfide, 97%
Sigma-Aldrich
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Allyl sulfide, ≥97%, FG
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Sigma-Aldrich
Sodium azide 0.1 M solution
Sigma-Aldrich
Acetaldehyde, puriss. p.a., anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Acetaldehyde, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Acetaldehyde, natural, FG
Sigma-Aldrich
Fomepizole, 99%
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Sigma-Aldrich
Alcohol Dehydrogenase from Saccharomyces cerevisiae, powder, ≥300 units/mg protein, mol wt ~141,000 (four subunits)
Sigma-Aldrich
Alcohol Dehydrogenase from Saccharomyces cerevisiae, ≥300 units/mg protein, lyophilized powder (contains buffer salts), Mw 141-151 kDa
Sigma-Aldrich
Alcohol Dehydrogenase from Saccharomyces cerevisiae
Sigma-Aldrich
4-Methylpyrazole hydrochloride, alcohol dehydrogenase inhibitor
Sigma-Aldrich
Alcohol Dehydrogenase equine, recombinant, expressed in E. coli, ≥0.5 U/mg
Supelco
Acetaldehyde, PESTANAL®, analytical standard