Merck
  • Cystamine ameliorates ventricular hypertrophy associated with modulation of IL-6-mediated signaling in lupus-prone mice.
Cystamine ameliorates ventricular hypertrophy associated with modulation of IL-6-mediated signaling in lupus-prone mice.
Life sciences (2013-02-13)
Bor-Show Tzang, Tsai-Ching Hsu, Tzy-Yen Chen, Chih-Yang Huang, Sin-Lun Li, Shao-Hsuan Kao
ABSTRACT
The aim of this study is to investigate the protective effects of cystamine on lupus-associated cardiac hypertrophy. Balb/c and lupus-prone NZB/W-F1 mice were individually randomized into sham group (saline, n=16) and cystamine group (n=16). Mice received saline or cystamine (100 mmol in 100 μL saline) by daily intraperitoneal injection for 2 consecutive weeks. Morphological, histological, and biochemical alterations were investigated. Cystamine decreased both left ventricular (LV) mass and LV mass/tissue-to-blood ratio (TBR) in NZB/W-F1 mice (p<0.05), whereas slight effects were observed in Balb/c mice. Moreover, cystamine reduced levels of atrial natriuretic peptide (ANP), C-reactive protein (CRP), heart type-fatty acid binding protein (h-FABP), creatine kinase-MB (CK-MB) and IL-6 in LV tissues of NZB/W-F1 mice (p<0.05). Additionally, in LV tissues of NZB/W-F1 mice, suppression of hypertrophic signaling mediated by IL-6 in response to administration of cystamine was revealed, including phosphorylation of MEK5, ERK5, c-Jun N-terminal kinase (JNK) and p38 mitogen-activated protein kinase (p38) (p<0.05). Cystamine alleviated LV hypertrophy in NZB/W-F1 mice as a result of decrease in hypertrophic mediators and suppression of IL-6 mediated hypertrophic signaling.
Sigma-Aldrich
Cystamine dihydrochloride, 96%
Sigma-Aldrich
Cystamine dihydrochloride, BioXtra
Sigma-Aldrich
Cystamine dihydrochloride, purum, ≥98.0% (AT)
Sigma-Aldrich
Transglutaminase from guinea pig liver, lyophilized powder, ≥1.5 units/mg protein