Merck
  • Adoptive transfer of human gingiva-derived mesenchymal stem cells ameliorates collagen-induced arthritis via suppression of Th1 and Th17 cells and enhancement of regulatory T cell differentiation.
Adoptive transfer of human gingiva-derived mesenchymal stem cells ameliorates collagen-induced arthritis via suppression of Th1 and Th17 cells and enhancement of regulatory T cell differentiation.
Arthritis and rheumatism (2013-02-13)
Maogen Chen, Wenru Su, Xiaohong Lin, Zhiyong Guo, Julie Wang, Qunzhou Zhang, David Brand, Bernhard Ryffel, Jiefu Huang, Zhongmin Liu, Xiaoshun He, Anh D Le, Song Guo Zheng
ABSTRACT
Current approaches offer no cures for rheumatoid arthritis (RA). Accumulating evidence has revealed that manipulation of bone marrow-derived mesenchymal stem cells (BM-MSCs) may have the potential to control or even prevent RA, but BM-MSC-based therapy faces many challenges, such as limited cell availability and reduced clinical feasibility. This study in mice with established collagen-induced arthritis (CIA) was undertaken to determine whether substitution of human gingiva-derived mesenchymal stem cells (G-MSCs) would significantly improve the therapeutic effects. CIA was induced in DBA/1J mice by immunization with type II collagen and Freund's complete adjuvant. G-MSCs were injected intravenously into the mice on day 14 after immunization. In some experiments, intraperitoneal injection of PC61 (anti-CD25 antibody) was used to deplete Treg cells in arthritic mice. Infusion of G-MSCs in DBA/1J mice with CIA significantly reduced the severity of arthritis, decreased the histopathology scores, and down-regulated the production of inflammatory cytokines (interferon-γ and interleukin-17A). Infusion of G-MSCs also resulted in increased levels of CD4+CD39+FoxP3+ cells in arthritic mice. These increases were noted early after infusion in the spleens and lymph nodes, and later after infusion in the synovial fluid. The FoxP3+ Treg cells that were increased in frequency mainly consisted of Helios-negative cells. When Treg cells were depleted, infusion of G-MSCs partially interfered with the progression of CIA. Pretreatment of G-MSCs with a CD39 or CD73 inhibitor significantly reversed the protective effect of G-MSCs on CIA. The role of G-MSCs in controlling the development and severity of CIA mostly depends on CD39/CD73 signals and partially depends on the induction of CD4+CD39+FoxP3+ Treg cells. G-MSCs provide a promising approach for the treatment of autoimmune diseases.
Sigma-Aldrich
5′-Nucleotidase human, recombinant, expressed in CHO cells, vial of 6-12 μg
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥60 units/mg protein, lyophilized powder (partially purified)
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥3.0 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder