Merck
  • Pancreatic lipase inhibitory activity of cassiamin A, a bianthraquinone from Cassia siamea.
Pancreatic lipase inhibitory activity of cassiamin A, a bianthraquinone from Cassia siamea.
Natural product communications (2013-03-22)
Dilip Kumar, Aniket Karmase, Sneha Jagtap, Ruchi Shekhar, Kamlesh K Bhutani
ABSTRACT
In continuation towards the discovery of potential antiobesity lead(s) from natural products, we have screened n-hexane, dichloromethane (DCM), ethyl acetate (EtOAc) and methanol (MeOH) extracts of 33 Indian medicinal plants (200 extracts) for in vitro pancreatic lipase inhibitory activity. Of the screened extracts, the EtOAc extract of Cassia siamea roots showed 74.3 +/- 1.4% enzyme inhibition at 250 microg/mL concentration. Bioassay guided fractionation of the active extract afforded 6 known compounds viz. chrysophanol (1), physcion (2), emodin (3), cassiamin A (4), friedelin (5) and cycloart-25-en-3beta,24-diol (6). These compounds were further evaluated for pancreatic lipase inhibitory activity. Cassiamin A (4), a bianthraquinone, was found to be most active with an IC50 value of 41.8 +/- 1.2 microM and compounds 2 and 5 were found to be moderate enzyme inhibitors. Results indicate the antiobesity potential of C. siamea through pancreatic lipase inhibition.
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, solution, ≥100,000 U/g, white, beige
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g