Merck
  • Lgr4-mediated Wnt/β-catenin signaling in peritubular myoid cells is essential for spermatogenesis.
Lgr4-mediated Wnt/β-catenin signaling in peritubular myoid cells is essential for spermatogenesis.
Development (Cambridge, England) (2013-03-28)
Yu Qian, Shijie Liu, Yuting Guan, Hongjie Pan, Xin Guan, Zhongwei Qiu, Liang Li, Na Gao, Yongxiang Zhao, Xiaoying Li, Yan Lu, Mingyao Liu, Dali Li
ABSTRACT
Peritubular myoid cells (PMCs) are myofibroblast-like cells that surround the seminiferous tubules and play essential roles in male fertility. How these cells modulate spermatogenesis and the signaling pathways that are involved are largely unknown. Here we report that Lgr4 is selectively expressed in mouse PMCs in the testes, and loss of Lgr4 leads to germ cells arresting at meiosis I and then undergoing apoptosis. In PMCs of Lgr4 mutant mice, the expression of androgen receptor, alpha-smooth muscle actin and extracellular matrix proteins was dramatically reduced. Malfunctioning PMCs further affected Sertoli cell nuclear localization and functional protein expression in Lgr4(-/-) mice. In addition, Wnt/β-catenin signaling was activated in wild-type PMCs but attenuated in those of Lgr4(-/-) mice. When Wnt/β-catenin signaling was reactivated by crossing with Apc(min/+) mice or by Gsk3β inhibitor treatment, the Lgr4 deficiency phenotype in testis was partially rescued. Together, these data demonstrate that Lgr4 signaling through Wnt/β-catenin regulates PMCs and is essential for spermatogenesis.
Sigma-Aldrich
β-Galactosidase from Escherichia coli, Grade VIII, lyophilized powder, ≥500 units/mg protein
Sigma-Aldrich
β-Galactosidase from Escherichia coli, Grade VI, lyophilized powder, ≥250 units/mg protein
Sigma-Aldrich
β-Galactosidase from Escherichia coli, aqueous glycerol suspension, ≥500 units/mg protein (biuret)
Sigma-Aldrich
β-Galactosidase from bovine liver, Grade III, lyophilized powder, ≥0.15 units/mg protein
Sigma-Aldrich
β-Galactosidase from Escherichia coli, lyophilized powder, ≥500 units/mg protein
Sigma-Aldrich
β-Galactosidase from bovine testes, ammonium sulfate suspension, 1.0-3.0 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
β-Galactosidase from Escherichia coli, suitable for enzyme immunoassay, lyophilized, powder, ~140 U/mg
Sigma-Aldrich
β-Galactosidase from Aspergillus oryzae, ≥8.0 units/mg solid