Merck
  • [The second generation universal oligonucleotide microarray for subtyping of influenza virus A].
[The second generation universal oligonucleotide microarray for subtyping of influenza virus A].
Bioorganicheskaia khimiia (2013-04-04)
E V Kostina, V A Riabinin, G A Maksakova, A N Siniakov
ABSTRACT
The microchip for influenza A subtyping was developed, functioning on a principle "one spot--one subtype". Each spot contains the set of oligonucleotide probes, specific for a particular subtype of hemagglutinin, neuraminidase or matrix gene. Reliability of the proposed chip version is the same as for earlier created in our group full-size microchip for separate hemagglutinin and neuraminidase subtyping. To visualize the image, analyzed DNA can be labeled by either fluorescent dye or biotin with the further fixation in system streptavidin-gold nanoparticles and image development by silver precipitation. In the second case common version of scanner can be used for the image analysis, that essentially simplifies procedure of influenza A subtyping.
Sigma-Aldrich
α(2→3) Neuraminidase from Streptococcus pneumoniae, buffered aqueous solution
Sigma-Aldrich
Neuraminidase Agarose from Clostridium perfringens (C. welchii), Type VI-A, ammonium sulfate suspension
Sigma-Aldrich
α(2→3,6) Neuraminidase from Clostridium perfringens (C. welchii), recombinant, expressed in E. coli, buffered aqueous solution, ≥250 units/mg protein
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type VIII, lyophilized powder, 10-20 units/mg protein (using 4MU-NANA), 3.5-8.0 units/mg protein (mucin)
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Suitable for manufacturing of diagnostic kits and reagents, Type V, lyophilized powder
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type X, lyophilized powder, ≥50 units/mg protein (using 4MU-NANA)
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type VI, lyophilized powder, 6-15 units/mg protein (using 4MU-NANA), 2-10 units/mg protein (mucin)
Sigma-Aldrich
α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, Proteomics Grade, suitable for MALDI-TOF MS
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, Type III, buffered aqueous solution, 0.2 μm filtered, 1-5 units/mg protein (Lowry, using NAN-lactose)
Sigma-Aldrich
α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, recombinant, expressed in E. coli, buffered aqueous solution
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, Type II, buffered aqueous solution, 8-24 units/mg protein (Lowry, using NAN-lactose)
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, ≥1.5 U/mL, specific activity ≥ 1.5U/mg protein