Merck
  • Vascular smooth muscle cells isolated from adipose triglyceride lipase-deficient mice exhibit distinct phenotype and phenotypic plasticity.
Vascular smooth muscle cells isolated from adipose triglyceride lipase-deficient mice exhibit distinct phenotype and phenotypic plasticity.
Biochemical and biophysical research communications (2013-04-16)
Yanhui Lin, Shunmei Chiba, Akira Suzuki, Satoshi Yamaguchi, Takaya Nakanishi, Hirofumi Matsumoto, Yoshihiko Ikeda, Hatsue Ishibashi-Ueda, Ken-ichi Hirano, Seiya Kato
ABSTRACT
The alteration of triglyceride (TG) metabolism in vascular smooth muscle cells (SMC) is likely to be correlated with certain phenotype, though this has not been elucidated. Adipose triglyceride lipase (ATGL) exerts major TG catalytic activity in both adipotic and non-adipotic cells. In the present study, we isolated SMC from ATGL-deficient mice (ATGL(-/-)mSMC). ATGL(-/-)mSMC showed spontaneous TG accumulation with lower mitogenic response and smooth muscle actin (SMA) expression compared to ATGL (+/+)mSMC. Percentage of senescence-associated β-galactosidase positive cells was also increased in ATGL(-/-)mSMC. Real-time PCR followed by screening with focused DNA array analysis revealed up-regulated expression of glucokinase (1.7-fold), lipoprotein lipase (3.8-fold) and interleukin-6 (3.7-fold) and down-regulated expression of vascular endothelial growth factor-A (0.2-fold), type I collagen (0.5-fold), and transforming growth factor-β (0.4-fold) in ATGL(-/-)mSMC compared to ATGL(+/+)mSMC. Next, ectopic gene transfer of human ATGL was attempted using doxycycline (Dox)-regulatable myc-DDK-tagged adenovirus vector (AdvATGL). AdvATGL infection resulted in a reduction of TG accumulation with elevated mitogenic response and SMA expression, and decreased in senescent cell numbers in ATGL(-/-)mSMC. Moreover, deviated gene expression pattern in ATGL(-/-)mSMC was potentially corrected. Our data suggest that ATGL(-/-)mSMC have a distinct phenotype that may be related to vascular pathogenesis. Plasticity of SMC phenotypes correlated to lipid metabolism could be a therapeutic target.
Sigma-Aldrich
Esterase from porcine liver, ammonium sulfate suspension, ≥150 units/mg protein (biuret)
Sigma-Aldrich
Esterase from rabbit liver, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Aspergillus oryzae, solution, ≥100,000 U/g, white, beige
Sigma-Aldrich
Esterase from porcine liver, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Esterase from porcine liver, lyophilized, powder, slightly beige, ≥50 U/mg
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Esterase Isoenzyme 1 porcine liver, recombinant, recombinant, expressed in E. coli, ≥30.0 U/g
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Esterase Pseudomonas fluorescens, recombinant from E. coli, ≥4 U/mg
Sigma-Aldrich
Carboxylesterase 2 human, recombinant, expressed in mouse NSO cells, ≥95% (SDS-PAGE)
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, ≥0.2 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg