Merck
Endonucleases.
Current protocols in molecular biology (2011-01-13)
Nicole M Nichols
ABSTRACT
Reaction conditions for a variety of endonucleases are detailed in this unit along with discussions of potential applications. Enzymes covered include BAL 31 nuclease, S1 nuclease, mung bean nuclease, micrococcal nuclease, and DNase I. A general discussion regarding the use of endonucleases to generate nonspecific breaks in dsDNA is also provided. For a detailed discussion of the endonucleases more typically associated with DNA damage repair (e.g., Endo III, IV, V and VIII of E. coli and human APE1), see UNIT 3.9.
Sigma-Aldrich
Nuclease S1 from Aspergillus oryzae, for single-strand DNA/RNA digestion
Sigma-Aldrich
Nuclease P1 from Penicillium citrinum, lyophilized powder, ≥200 units/mg protein (E1%/280, 3′-5′-Phosphodiesterase)