Merck
  • [Influence of ethanol-metabolising systems on intensity of lipid peroxidation processes in gastrointestinal tract of rats].
[Influence of ethanol-metabolising systems on intensity of lipid peroxidation processes in gastrointestinal tract of rats].
Biomeditsinskaia khimiia (2013-05-09)
N É Petushok, T Ch Grokhovskaia, N G Mel'nichenko, S P Pron'ko
ABSTRACT
The effects of some ethanol-metabolising systems (aldehyde dehydrogenase, catalase, cytochrome P450 2E1) on activation of lipid peroxidation (LPO) processes in gastrointestinal tract of rats have been studied using inhibitors of these systems. The intensity of LPO processes was evaluated by thiobarbituric acid-reactive products and chemiluminiscence intensity. It was found, that the acetadehyde metabolism play the main role in the indiction of lipid peroxidation in the gastrointestinal tract of rats.
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Sigma-Aldrich
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Acetaldehyde, natural, FG
Sigma-Aldrich
Acetaldehyde, puriss. p.a., anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Acetaldehyde, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Sigma-Aldrich
Catalase from Corynebacterium glutamicum, solution, deep brown, ≥500000 U/mL
Sigma-Aldrich
Catalase from bovine liver, powder, suitable for cell culture, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from human erythrocytes, ≥90% (SDS-PAGE), buffered aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, ≥10,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 40,000-60,000 units/mg protein (E1%/405)
Sigma-Aldrich
Catalase from Aspergillus niger, ammonium sulfate suspension, ≥4,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 10,000-40,000 units/mg protein
Supelco
Acetaldehyde, PESTANAL®, analytical standard