Merck
  • Repeated fermentation from raw starch using Saccharomyces cerevisiae displaying both glucoamylase and α-amylase.
Repeated fermentation from raw starch using Saccharomyces cerevisiae displaying both glucoamylase and α-amylase.
Enzyme and microbial technology (2012-04-17)
Syun-ichi Yamakawa, Ryosuke Yamada, Tsutomu Tanaka, Chiaki Ogino, Akihiko Kondo
ABSTRACT
A diploid yeast strain displaying both α-amylase and glucoamylase was developed for repeated fermentation from raw starch. First, the construct of α-amylase was optimized for cell surface display, as there have been no reports of α-amylase-displaying yeast. The modified yeast displaying both glucoamylase and α-amylase produced 46.5 g/l of ethanol from 200 g/l of raw corn starch after 120 h of fermentation, and this was 1.5-fold higher when compared to native α-amylase-displaying yeast. Using the glucoamylase and modified α-amylase co-displaying diploid strain, we repeated fermentation from 100g/l of raw starch for 23 cycles without the loss of α-amylase or glucoamylase activity. The average ethanol productivity and yield during repeated fermentation were 1.61 g/l/h and 76.6% of the theoretical yield, respectively. This novel yeast may be useful for reducing the cost of bio-ethanol production and may be suitable for industrial-scale bio-ethanol production.
Sigma-Aldrich
Amyloglucosidase from Aspergillus niger, lyophilized powder, 30-60 units/mg protein (biuret), ≤0.02% glucose
Sigma-Aldrich
Amyloglucosidase from Aspergillus niger, Isoelectric focusing marker, pI 3.6
Sigma-Aldrich
Amyloglucosidase from Aspergillus niger, ≥260 U/mL, aqueous solution
Sigma-Aldrich
Amyloglucosidase from Aspergillus niger, powder, white, ~120 U/mg
Sigma-Aldrich
Amyloglucosidase from Aspergillus niger, lyophilized, powder, ~70 U/mg
Sigma-Aldrich
Amyloglucosidase from Aspergillus niger, ammonium sulfate suspension, ≥40 units/mg protein
Sigma-Aldrich
Amyloglucosidase from Rhizopus sp., ≥40,000 units/g solid