Merck
  • The adipose tissue triglyceride lipase ATGL/PNPLA2 is downregulated by insulin and TNF-alpha in 3T3-L1 adipocytes and is a target for transactivation by PPARgamma.
The adipose tissue triglyceride lipase ATGL/PNPLA2 is downregulated by insulin and TNF-alpha in 3T3-L1 adipocytes and is a target for transactivation by PPARgamma.
American journal of physiology. Endocrinology and metabolism (2006-05-18)
Ji Young Kim, Kristin Tillison, Jun-Ho Lee, David A Rearick, Cynthia M Smas
ABSTRACT
The minimal adipose phenotype of hormone-sensitive lipase (HSL)-null mice suggested that other hormonally responsive lipase(s) were present in adipocytes. Recent studies have characterized a new adipose tissue triglyceride lipase, ATGL/PNPLA2/destnutrin/iPLA2zeta/TTS2.2 (ATGL). We had previously cloned a novel adipose-enriched transcript by differential screening and recently determined its identity with murine ATGL. We report here on the regulation of ATGL by TNF-alpha and insulin in 3T3-L1 adipocytes and identify ATGL as a target for transcriptional activation by the key adipogenic transcription factor PPARgamma. Insulin at 100 nM resulted in a marked decrease in ATGL transcript that was effectively blocked by inhibitors for PI 3-kinase and p70 ribosomal protein S6 kinase. TNF-alpha treatment decreased ATGL transcript in a time-dependent manner that paralleled TNF-alpha downregulation of PPARgamma with a maximal decrease noted by 6 h. TNF-alpha effects on ATGL were attenuated by pretreatment with PD-98059, LY-294002, or rapamycin, suggesting involvement of the p44/42 MAP kinase, PI 3-kinase, and p70 ribosomal protein S6 kinase signals. To study transcriptional regulation of ATGL, we cloned 2,979 bp of the murine ATGL 5'-flanking region. Compared with promoterless pGL2-Basic, the -2979/+21 ATGL luciferase construct demonstrated 120- and 40-fold increases in activity in white and brown adipocytes, respectively. Luciferase reporter activities for a series of eight ATGL promoter deletions revealed that the -928/+21, -1738/+21, -1979/+21, and -2979/+21 constructs were transactivated by PPARgamma. Our findings identify the novel lipase ATGL to be a target gene for TNF-alpha and insulin action in adipocytes and reveal that it is subject to transcriptional control by PPARgamma-mediated signals.
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
PD 98,059, solid
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)