Merck
  • cDNA cloning of human-milk bile-salt-stimulated lipase and evidence for its identity to pancreatic carboxylic ester hydrolase.
cDNA cloning of human-milk bile-salt-stimulated lipase and evidence for its identity to pancreatic carboxylic ester hydrolase.
European journal of biochemistry (1990-09-11)
J Nilsson, L Bläckberg, P Carlsson, S Enerbäck, O Hernell, G Bjursell
ABSTRACT
We have isolated and sequenced cDNA clones covering the entire coding sequence of human-milk bile-salt-stimulated lipase, as well as 996 nucleotides of the 3' end of the pancreatic enzyme carboxylic ester hydrolase. The deduced amino acid sequence of the lipase starts with a 23-residue leader peptide. The open reading frame continues with 722 amino acid residues. The sequence contains in the C-terminal part a proline-rich repeat, 16 repeats of 11 amino acid residues each. The mRNA was estimated to be approximately 2500 nucleotides from Northern blot and of similar size in mammary and pancreatic tissues. Data obtained indicate that the lipase and the carboxylesterase are identical and coded for by the same gene. The cDNA is 2428 bases long, which indicates that a near full-length copy of the transcript has been isolated. Comparisons with other enzymes show that the lipase is a new member of the supergene family of serine hydrolases. It is not only closely related (and in its N-terminal half virtually identical) to lysophospholipase from rat pancreas and cholesterol esterase from bovine pancreas, but also shows a high degree of similarity to several esterases, e.g. acetylcholine esterase. In contrast, no such similarity could be found to typical lipases.
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid