Merck
  • The -250G>A promoter variant in hepatic lipase associates with elevated fasting serum high-density lipoprotein cholesterol modulated by interaction with physical activity in a study of 16,156 Danish subjects.
The -250G>A promoter variant in hepatic lipase associates with elevated fasting serum high-density lipoprotein cholesterol modulated by interaction with physical activity in a study of 16,156 Danish subjects.
The Journal of clinical endocrinology and metabolism (2008-03-28)
Niels Grarup, Camilla H Andreasen, Mette K Andersen, Anders Albrechtsen, Annelli Sandbaek, Torsten Lauritzen, Knut Borch-Johnsen, Torben Jørgensen, Ole Schmitz, Torben Hansen, Oluf Pedersen
ABSTRACT
Hepatic lipase plays a pivotal role in the metabolism of high-density lipoprotein (HDL) and low-density lipoprotein by involvement in reverse cholesterol transport and the formation of atherogenic small dense low-density lipoprotein. The objective was to investigate the impact of variants in LIPC on metabolic traits and type 2 diabetes in a large sample of Danes. Because behavioral factors influence hepatic lipase activity, we furthermore examined possible gene-environment interactions in the population-based Inter99 study. The LIPC -250G>A (rs2070895) variant was genotyped in the Inter99 study (n = 6070), the Anglo-Danish-Dutch Study of Intensive Treatment in People with Screen Detected Diabetes in Primary Care Denmark screening cohort of individuals with risk factors for undiagnosed type 2 diabetes (n = 8662), and in additional type 2 diabetic patients (n = 1,064) and glucose-tolerant control subjects (n = 360). In the Inter99 study, the A allele of rs2070895 associated with a 0.057 mmol/liter [95% confidence interval (CI) 0.039-0.075] increase in fasting serum HDL-cholesterol (HDL-c) (P = 8 x 10(-10)) supported by association in the Anglo-Danish-Dutch Study of Intensive Treatment in People with Screen Detected Diabetes in Primary Care study [0.038 mmol/liter per allele (95% CI 0.024-0.053); P = 2 x 10(-7)). The allelic effect on HDL-c was modulated by interaction with self-reported physical activity (P(interaction) = 0.002) because vigorous physically active homozygous A-allele carriers had a 0.30 mmol/liter (95% CI 0.22-0.37) increase in HDL-c compared with homozygous G-allele carriers. We validate the association of LIPC promoter variation with fasting serum HDL-c and present data supporting an interaction with physical activity implying an increased effect on HDL-c in vigorous physically active subjects carrying the -250 A allele. This interaction may have potential implications for public health and disease prevention.
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)