Merck
  • Polymorphisms in the adiponutrin gene are associated with increased insulin secretion and obesity.
Polymorphisms in the adiponutrin gene are associated with increased insulin secretion and obesity.
European journal of endocrinology (2008-08-30)
Lovisa E Johansson, Ulf Lindblad, Charlotte A Larsson, Lennart Råstam, Martin Ridderstråle
ABSTRACT
The insulin responsive adiponutrin or patatin-like phospholipase 3 (PNPLA3, previously ADPN) gene shows association with obesity and in vitro adipocyte lipolysis. This study aimed to replicate the association between PNPLA3 variants and obesity, and to investigate their effect on insulin resistance and beta-cell function. rs738409 (Met148Ile) and rs2072907 (C to G) were genotyped using TaqMan allelic discrimination assay in a Swedish population-based sample (n=1811). Oral glucose tolerance test (OGTT) with data from three time points (0, 30, and 120 min) were available from individuals under the age of 50 years (n=973). Both variant alleles were associated with decreased prevalence of obesity (P<0.05); odds ratio 0.75 (0.61-0.93) per carried Ile-allele for rs738409 and 0.80 (0.64-1.00) per carried G-allele for rs2072907. For obesity as a quantitative trait, there was no association in the whole population, but in obese subjects body mass index (BMI; P=0.023) and waist (P=0.0098) were higher in carriers of the Ile-allele. The Ile-carriers also displayed decreased insulin secretion in response to OGTT (30 min insulin; P=0.007, insulinogenic index; P=0.0051) with no significant differences in fasting plasma glucose (P=0.31), beta-cell function (disposition index; P=0.17) or homeostasis model of assessment insulin resistance (HOMA-IR; P=0.063). The correlation between BMI and HOMA-IR differed (Met/X versus Ile/Ile, P=0.028), Met-allele carriers were seemingly more insulin resistant at a lower BMI. The rs2072907 variant shows similar results for insulin secretion. The significance of this finding remained after adjusting for age, gender, and level of self-reported leisure-time physical activity. We confirm the association between PNPLA3 and obesity. In addition, the rs738409 variant was associated with insulin secretion. There seems to be a differential effect of the Ile-allele depending on the degree of obesity, possibly as a consequence of insulin resistance.
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)