Merck
Identification of G protein alpha subunit-palmitoylating enzyme.
Molecular and cellular biology (2008-11-13)
Ryouhei Tsutsumi, Yuko Fukata, Jun Noritake, Tsuyoshi Iwanaga, Franck Perez, Masaki Fukata
ABSTRACT
The heterotrimeric G protein alpha subunit (Galpha) is targeted to the cytoplasmic face of the plasma membrane through reversible lipid palmitoylation and relays signals from G-protein-coupled receptors (GPCRs) to its effectors. By screening 23 DHHC motif (Asp-His-His-Cys) palmitoyl acyl-transferases, we identified DHHC3 and DHHC7 as Galpha palmitoylating enzymes. DHHC3 and DHHC7 robustly palmitoylated Galpha(q), Galpha(s), and Galpha(i2) in HEK293T cells. Knockdown of DHHC3 and DHHC7 decreased Galpha(q/11) palmitoylation and relocalized it from the plasma membrane into the cytoplasm. Photoconversion analysis revealed that Galpha(q) rapidly shuttles between the plasma membrane and the Golgi apparatus, where DHHC3 specifically localizes. Fluorescence recovery after photobleaching studies showed that DHHC3 and DHHC7 are necessary for this continuous Galpha(q) shuttling. Furthermore, DHHC3 and DHHC7 knockdown blocked the alpha(1A)-adrenergic receptor/Galpha(q/11)-mediated signaling pathway. Together, our findings revealed that DHHC3 and DHHC7 regulate GPCR-mediated signal transduction by controlling Galpha localization to the plasma membrane.
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Sigma-Aldrich
2-Bromohexadecanoic acid, ≥99.0% (GC)
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
2-Bromohexadecanoic acid, ~97%
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Prazosin hydrochloride, ≥99.0% (HPLC)