Merck
  • A nonsynonymous gene variant in the adiponutrin gene is associated with nonalcoholic fatty liver disease severity.
A nonsynonymous gene variant in the adiponutrin gene is associated with nonalcoholic fatty liver disease severity.
Journal of lipid research (2009-09-10)
Silvia Sookoian, Gustavo O Castaño, Adriana L Burgueño, Tomas Fernández Gianotti, María Soledad Rosselli, Carlos Jose Pirola
ABSTRACT
We explored the role of the adiponutrin (PNPLA3) nonsynonymous-rs738409 single nucleotide polymorphism (SNP) in genetic susceptibility to nonalcoholic fatty liver disease (NAFLD) and whether this SNP contributes to the severity of histological disease. Two hundred sixty-six individuals were evaluated in a case-control association study, which included 172 patients with features of NAFLD and 94 control subjects. The rs738409 G allele was significantly associated with NAFLD (P < 0.001; OR 2.8 95%, CI 1.5-5.2), independent of age, sex, body mass index (BMI), and Homeostasis Model Assessment (HOMA) index. When we tested the hypothesis of a relation between the SNP and the histological spectrum of NAFLD, a significant association was observed [chi2 19.9, degree of freedom (df): 2, P < 5 x 10(-5), adjusted for HOMA and BMI]. The degree of liver steatosis, as evaluated by liver biopsy, was significantly associated with the rs738409 G allele. Patients with CC genotype showed a lower steatosis score (14.9% +/- 3.9) in comparison with the CG genotype (26.3% +/- 3.5) and GG genotype (33.3% +/- 4.0) (P < 0.005). The proportion of the total variation attributed to rs738409 genotypes was 5.3% (beta 0.23 +/- 0.07; P < 0.002). Our data suggest that the rs738409 G allele is associated not only with fat accumulation in the liver but also with liver injury, possibly triggered by lipotoxicity.
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger