Merck
  • Isolation and cDNA sequence of human postheparin plasma hepatic triglyceride lipase.
Isolation and cDNA sequence of human postheparin plasma hepatic triglyceride lipase.
The Journal of biological chemistry (1988-08-05)
G A Martin, S J Busch, G D Meredith, A D Cardin, D T Blankenship, S J Mao, A E Rechtin, C W Woods, M M Racke, M P Schafer
ABSTRACT
Hepatic triglyceride lipase (H-TGL) was isolated from human postheparin plasma by column chromatography on heparin-Sepharose and phenyl-Sepharose and immunoaffinity chromatography with monoclonal antibodies. The purified enzyme had an apparent molecular weight of 65,000 on sodium dodecyl sulfate-polyacrylamide gel electrophoresis and an amino-terminal sequence of Leu-Gly-Gln-Ser-Leu-Lys-Pro-Glu. Partial amino acid sequences of seven cyanogen bromide peptides were obtained. A human hepatoma cDNA library was screened with synthetic oligonucleotides derived from the partial protein sequence. The cloned H-TGL cDNA of 1569 nucleotides predicts a mature protein of 477 amino acids plus a leader sequence of 22 amino acids. Blot hybridization analysis of poly(A)+ mRNA with a putative H-TGL cDNA clone gave a single hybridizing band of 1.7 kilobases. The protein contains four consensus N-glycosylation sequences based on the cDNA sequence. Comparison of the enzyme sequence with that of other lipases reveals highly conserved sequences in regions of putative lipid and heparin binding. The carboxyl terminus of H-TGL contains a highly basic sequence which is not reported to be present in rat H-TGL or other members of the lipase gene family.
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g