Merck
  • Molecular cloning of a human gastric lipase and expression of the enzyme in yeast.
Molecular cloning of a human gastric lipase and expression of the enzyme in yeast.
Biochimica et biophysica acta (1987-08-25)
M W Bodmer, S Angal, G T Yarranton, T J Harris, A Lyons, D J King, G Pieroni, C Riviere, R Verger, P A Lowe
ABSTRACT
The molecular cloning of a cDNA coding for human gastric lipase and its expression in yeast is described. A lipase present in human gastric aspirates was purified and its N-terminal amino-acid sequence was determined. This was found to be homologous with the N-terminal sequence of rat lingual lipase. A cDNA library was constructed from mRNA isolated from human stomach tissue and probed with cloned rat lingual lipase DNA. One clone, pGL17, consisting of approximately 1450 base-pairs, contained the entire coding sequence for a human gastric lipase. The amino-acid sequence from the isolated protein and the DNA sequence obtained from the cloned gene indicated that human gastric lipase consists of a 379 amino acid polypeptide with an unglycosylated Mr of 43,162. Human gastric lipase and rat lingual lipase amino-acid sequences were closely homologous but were unrelated to porcine pancreatic lipase apart from a 6 amino-acid sequence around the essential Ser-152 of porcine pancreatic lipase. A yeast expression plasmid containing the phosphoglycerate kinase promoter and terminator sequences together with the human gastric lipase gene was constructed. Yeast transformed with this vector synthesised the lipolytically active enzyme.
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg