Merck
  • Human small-intestinal beta-galactosidases. Separation and characterization of one lactase and one hetero beta-galactosidase.
Human small-intestinal beta-galactosidases. Separation and characterization of one lactase and one hetero beta-galactosidase.
The Biochemical journal (1969-09-01)
N G Asp, A Dahlqvist, O Koldovský
ABSTRACT
1. Two beta-galactosidases from human small-intestinal mucosa were separated by gel-filtration chromatography and the properties of the two enzymes were studied. Lactose and four hetero beta-galactosides were used as substrates. 2. One of the enzymes was particle-bound and could be partially solubilized with papain. Of the substrates hydrolysed by this enzyme, lactose was hydrolysed most rapidly. This enzyme is thus essentially a disaccharidase and is named lactase. It is presumably identical with the ;lactase 1' described earlier. 3. The other enzyme was mainly soluble and hydrolysed all artificial substrates used, whereas no lactase activity could be detected. This enzyme has therefore been designated hetero beta-galactosidase. 4. p-Chloromercuribenzoate (0.1mm) inhibited the hetero beta-galactosidase completely but did not influence the activity of the lactase. Tris was a competitive inhibitor of both enzymes. 5. The residual lactase activity in the mucosa of lactose-intolerant patients may be exerted by a small amount of remaining lactase as such, or possibly by a third enzyme with a more acid pH optimum.
Sigma-Aldrich
β-Galactosidase from Aspergillus oryzae, ≥8.0 units/mg solid
Sigma-Aldrich
β-Galactosidase from Escherichia coli, suitable for enzyme immunoassay, lyophilized, powder, ~140 U/mg
Sigma-Aldrich
β-Galactosidase from Escherichia coli, aqueous glycerol suspension, ≥500 units/mg protein (biuret)
Sigma-Aldrich
β-Galactosidase from bovine testes, ammonium sulfate suspension, 1.0-3.0 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
β-Galactosidase from bovine liver, Grade III, lyophilized powder, ≥0.15 units/mg protein
Sigma-Aldrich
β-Galactosidase from Escherichia coli, Grade VI, lyophilized powder, ≥250 units/mg protein
Sigma-Aldrich
β-Galactosidase from Escherichia coli, Grade VIII, lyophilized powder, ≥500 units/mg protein
Sigma-Aldrich
β-Galactosidase from Escherichia coli, lyophilized powder, ≥500 units/mg protein