Merck
  • Role of intrarenal renin-angiotensin system on pressure-natriuresis in spontaneously hypertensive rats.
Role of intrarenal renin-angiotensin system on pressure-natriuresis in spontaneously hypertensive rats.
Clinical and Experimental Hypertension (1990)
Takenaka, T.