Merck
  • Cholinergic and glutamatergic transmission in medial vestibular nucleus neurons responding to lateral roll tilt in rats.
Cholinergic and glutamatergic transmission in medial vestibular nucleus neurons responding to lateral roll tilt in rats.
Brain research (1999-10-13)
S Takeshita, M Sasa, K Ishihara, H Matsubayashi, K Yajin, M Okada, R Izumi, K Arita, K Kurisu
ABSTRACT
The responses of the medial vestibular nucleus (MVN) neurons to lateral tilt and the neurotransmitters mediating otolith information to MVN neurons were investigated using rats. A computer-operated goniometer was tilted 20 degrees clockwise and counterclockwise at an angular speed of 5 degrees /s and paused in the inclined positions for 10 s to record neuronal responses in the static phase. The 185 MVN neurons recorded were classified into eight types according to their responses to tilt (alpha, beta, gamma, delta, epsilon, zeta, eta and theta). A majority showed increased firing in response to ipsilateral tilting and decreased firing in response to contralateral tilting (alpha type: 31.4%) or exhibited the reverse pattern (beta type: 36.8%). Further, other groups of neurons increased (gamma type) or decreased (delta type) firing rates to either side tilting and increased (epsilon and zeta type) or decreased (eta and theta type) firing only on one side. Atropine or L-glutamic acid diethyl ester hydrochloride (GDEE) applied microiontophoretically antagonized tilt-induced firing of alpha type neurons in 58.8% or 60.0%, respectively, and of beta type neurons in 66.7% or 58.3%, respectively. When the effects of atropine and GDEE were examined in the same neurons, antagonizing effects of both drugs on tilt-induced firing were obtained in 28.6% and 40.0% of alpha and beta type neurons, respectively. These results suggest that both acetylcholine and glutamate act as neurotransmitters in the transmission of otolith information to most MVN neurons.
Sigma-Aldrich
L-Glutamic acid diethyl ester hydrochloride, 97%