Merck
Odour-impact compounds of Gorgonzola cheese.
The Journal of dairy research (2000-06-07)
L Moio, P Piombino, F Addeo
ABSTRACT
Volatile concentrates were obtained by vacuum distillation from both natural and creamy Gorgonzola cheese and isolated by continuous liquid-liquid extraction. Both were analysed by high resolution gas chromatography (HRGC), HRGC-mass spectrometry and HRGC-olfactometry. A total of 63 components were identified in the neutral extract of the natural type (21 esters, 13 ketones, 14 alcohols, 5 aldehydes, 1 sulphur compound, 7 aromatic compounds and 2 terpenes) and 52 in the creamy type (17 esters, 12 ketones, 10 alcohols, 5 aldehydes, 1 sulphur compound, 5 aromatic compounds and 2 terpenes). Ketones, whose major components were 2-nonanone and 2-heptanone, were the predominant constituents of the neutral fraction. By olfactometric analysis of the neutral extracts, 23 odour-impact compounds were found in the natural and 21 in the creamy Gorgonzola cheese. 1-Octen-3-ol, ethyl hexanoate, 2-nonanone, 2-heptanone, 2-heptanol, ethyl butanoate, 2-nonanol and 4-methylanisole were the key odorants of the natural cheese, whereas 2-heptanone, 2-heptanol, ethyl butanoate, 3-methyl thiopropanal and an unidentified constituent with a fruity odour were characteristic of the creamy Gorgonzola cheese. On the basis of high odour unity values, 2-nonanone, 1-octen-3-ol, 2-heptanol, ethyl hexanoate, methylanisole and 2-heptanone were the most important odorants of natural and creamy Gorgonzola cheese aroma.
Sigma-Aldrich
2-Nonanone, ≥99%
Sigma-Aldrich
2-Nonanone, natural, ≥97%, FCC, FG
Sigma-Aldrich
2-Nonanone, ≥99%, FCC, FG
Sigma-Aldrich
1,3-Dimethoxybenzene, ≥98%, FG