Merck
  • Solid-phase synthesis of a modified 13-residue seminalplasmin fragment on 1,6-hexanediol diacrylate-crosslinked polystyrene support.
Solid-phase synthesis of a modified 13-residue seminalplasmin fragment on 1,6-hexanediol diacrylate-crosslinked polystyrene support.
Peptides (2000-08-26)
C Arunan, R Nagaraj, V N Rajasekharan Pillai
ABSTRACT
A novel 1,6-hexanediol diacrylate cross-linked resin was prepared that was subsequently functionalized by using chloromethyl methyl ether to afford a high-capacity resin. The resin exhibited good swelling and its application in the successful synthesis of a 13-residue peptide corresponding to the fragment of seminalplasmin has been illustrated. The resin was chemically inert at peptide synthetic conditions.
Sigma-Aldrich
1,6-Hexanediol solution, BioUltra, ~6 M in H2O
Sigma-Aldrich
1,6-Hexanediol, 99%
Sigma-Aldrich
1,6-Hexanediol diacrylate, technical grade, 80%
Sigma-Aldrich
1,6-Hexanediol, 97%