Merck
  • The use of silylation for minimizing the interference effects caused by contamination in ultra-low level mercury analytics.
The use of silylation for minimizing the interference effects caused by contamination in ultra-low level mercury analytics.
Analytical and bioanalytical chemistry (2002-05-16)
Teemu Näykki
ABSTRACT
The use of silylation of glassware in mercury analytics was investigated. By treating the glassware with 5% (v/v) dichlorodimethylsilane in toluene, the surfaces of the vessels were inactivated. Method development proved that silylation can extend the range of stabile, contamination-free area as low as 1 ng/L mercury without any significant investments in laboratory clean-room facilities. The resilylation had to be done once a month.
Sigma-Aldrich
Dichlorodimethylsilane, ≥98.5% (GC)
Sigma-Aldrich
Dichlorodimethylsilane, ≥99.5%
Sigma-Aldrich
Dichlorodimethylsilane, Wacker Chemie AG, ≥99.0% (GC)
Supelco
Silanization solution I, ~5% (dimethyldichlorosilane in heptane), Selectophore