Merck
  • Stereospecific synthesis of polyfunctionalized carbacephams induced by titanocene(III) chloride.
Stereospecific synthesis of polyfunctionalized carbacephams induced by titanocene(III) chloride.
The Journal of organic chemistry (2003-03-01)
Gema Ruano, Justo Martiáñez, Manuel Grande, Josefa Anaya
ABSTRACT
Enantiomerically pure N-substituted epoxyalkene-2-azetidinones reacted with titanocene monochloride to give stereospecifically polyfunctionalized bicyclic beta-lactams. Four isomeric epoxyaldehydes 2 reacted with TiCp2Cl to give exclusively the respective carbacephams 7 while under the same reaction conditions the epoxyesters 1, which are more hindered for an intramolecular addition, gave the cyclization products 6 (only two isomers) and/or the elimination products 5 (all isomers).
Sigma-Aldrich
Titanium(III) chloride, ≥99.995% trace metals basis
Sigma-Aldrich
Titanium(III) chloride solution, 10-15% TiCl3 basis