Merck
  • High cerebral scyllo-inositol: a new marker of brain metabolism disturbances induced by chronic alcoholism.
High cerebral scyllo-inositol: a new marker of brain metabolism disturbances induced by chronic alcoholism.
Magma (New York, N.Y.) (2004-09-02)
A Viola, F Nicoli, B Denis, S Confort-Gouny, Y Le Fur, J-P Ranjeva, P Viout, P J Cozzone
ABSTRACT
Cerebral metabolic changes that concur to motor and/or cognitive disorders in actively drinking alcoholics are not well established. We tested the hypothesis that chronic alcoholics exhibit profound alterations in the cerebral metabolism of scyllo-inositol. Brain metabolism was explored in nine actively drinking and 11 recently detoxified chronic alcoholics by in vivo brain (1)H-MRS and in vitro(1)H-MRS of blood serum and cerebrospinal fluid. The cohort was composed of individuals with acute, subacute or chronic encephalopathy or without any clinical encephalopathy. Chronic alcoholism is associated with a hitherto unrecognized accumulation of brain scyllo-inositol. Our results suggest that scyllo-inositol is produced within the central nervous system and shows a diffuse but heterogenous distribution in brain where it can persist several weeks after detoxification. Its highest levels were observed in subjects with a clinically symptomatic alcohol-related encephalopathy. When detected, brain scyllo-inositol takes part in a metabolic encephalopathy since it is associated with reduced N-acetylaspartate and increased creatine. High levels of cerebral scyllo-inositol are correlated with altered glial and neuronal metabolism. Our findings suggest that the accumulation of scyllo-inositol may precede and take part in the development of symptomatic alcoholic metabolic encephalopathy.
Product Description
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Nicotinic acid, analytical standard
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Sigma-Aldrich
Nicotinic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Nicotinic acid, meets USP testing specifications
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
scyllo-Inositol, ≥98%
Sigma-Aldrich
Nicotinic acid, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinic acid, ≥98%
Supelco
Niacin (Nicotinic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture