Merck
  • Kinetics and thermodynamics of RRF, EF-G, and thiostrepton interaction on the Escherichia coli ribosome.
Kinetics and thermodynamics of RRF, EF-G, and thiostrepton interaction on the Escherichia coli ribosome.
Biochemistry (2004-10-06)
Hyuk-Soo Seo, Michael Kiel, Dongli Pan, V Samuel Raj, Akira Kaji, Barry S Cooperman
ABSTRACT
Ribosome recycling factor (RRF) and elongation factor-G (EF-G) are jointly essential for recycling bacterial ribosomes following termination of protein synthesis. Here we present equilibrium and rapid kinetic measurements permitting formulation of a minimal kinetic scheme that accounts quantitatively for RRF and EF-G interaction on the Escherichia coli ribosome. RRF and EF-G (a) each form a binary complex on binding to a bare ribosome which undergoes isomerization to a more stable complex, (b) form mixed ternary complexes on the ribosome in which the affinity for each factor is considerably lower than its affinity for binding to a bare ribosome, and (c) each bind to two sites per ribosome, with EF-G having considerably higher second-site affinity than RRF. Addition of EF-G to the ribosome-RRF complex induces rapid RRF dissociation, at a rate compatible with the rate of ribosome recycling in vivo, but added RRF does not increase the lability of ribosome-bound EF-G. Added thiostrepton slows the initial binding of EF-G, and prevents both formation of the more stable EF-G complex and EF-G-induced RRF dissociation. These findings are relevant for the mechanism of post-termination complex disassembly.
Sigma-Aldrich
7-Diethylamino-3-(4-maleimidophenyl)-4-methylcoumarin, ≥95% (HPLC), solid
Sigma-Aldrich
7-Diethylamino-3-(4-maleimidophenyl)-4-methylcoumarin, suitable for fluorescence, BioReagent, ≥90% (HPCE)