Merck
Synthesis, structure and pharmacology of acyl-2,6-xylidines.
Acta poloniae pharmaceutica (2004-10-16)
Włodzimierz Białasiewicz, Wincenty Kwapiszewski, Elzbieta Zurek, Andrzej Olczak, Marek Główka, Rościsław Polakowski
ABSTRACT
L-2-perhydroheterocyclicalkyl acids were condensed with 2,6-xylidine. 8 new optically active acyl-2,6-xylidines were obtained. Absolute configuration of acyl-2,6-xylidines were selected for pharmacological examinations.
Sigma-Aldrich
Tocainide hydrochloride, ≥98% (HPLC), solid
Supelco
2,6-Dimethylaniline, PESTANAL®, analytical standard
Sigma-Aldrich
2,6-Dimethylaniline, 99%
Supelco
2,6-Dimethylaniline, analytical standard