Merck
  • Redetermination of the extinction coefficient of camphor-10-sulfonic acid, a calibration standard for circular dichroism spectroscopy.
Redetermination of the extinction coefficient of camphor-10-sulfonic acid, a calibration standard for circular dichroism spectroscopy.
Analytical biochemistry (2004-11-24)
Andrew J Miles, Frank Wien, B A Wallace
Sigma-Aldrich
(1R)-(−)-10-Camphorsulfonic acid, 98%
Sigma-Aldrich
Camphor-10-sulfonic acid (β), 98%
Sigma-Aldrich
(−)-Camphor-10-sulfonic acid, purum, ≥98.0% (T)
Sigma-Aldrich
(+)-Camphor-10-sulfonic acid, purum, ≥98.0% (T)
Sigma-Aldrich
(1S)-(+)-10-Camphorsulfonic acid, 99%