Merck
[Preparation and spectral properties of PVP-modified CdS nanorods].
Guang pu xue yu guang pu fen xi = Guang pu (2005-10-06)
Qiu-li Yuan, Qiu-lin Nie, Wei-xiang Chen, Zhu-de Xu
ABSTRACT
Polyvinylpyrrolidone (PVP)-modified CdS nanorods were prepared by a hydrothermal reaction of CdCl2 2.5H2O and (NH4)2S with 10 wt% ethylenediamine aqueous solution as solvent and 1.0 wt% PVP as additives. The obtained products were characterized by means of XRD, TEM, IR, DTA-TG, UV-Vis and PL spectroscopies. The surface-modified CdS nanorods showed narrow size distribution and enhanced luminescence property compared with that of the unmodified ones. The UV-Vis spectra exhibited a structure peak. These results were attributed to the surface passivation of the CdS nanorods by the PVP molecules.
Sigma-Aldrich
Ammonium sulfide solution, 20 wt. % in H2O
Sigma-Aldrich
Ammonium sulfide solution, 20% in H2O
Sigma-Aldrich
Ammonium sulfide solution, 40-48 wt. % in H2O