Merck
  • [Mechanisms of peroxidase oxidation of o-dianisidine, 3,3',5,5'-tetramethylbenzidine, and o-phenylenediamine in the presence of sodium dodecyl sulfate].
[Mechanisms of peroxidase oxidation of o-dianisidine, 3,3',5,5'-tetramethylbenzidine, and o-phenylenediamine in the presence of sodium dodecyl sulfate].
Bioorganicheskaia khimiia (2006-03-10)
A V Kireĭko, I A Veselova, T N Shekhovtsova
ABSTRACT
Peroxidase oxidation of o-dianisidine, 3,3',5,5'-tetramethylbenzidine, and o-phenylenediamine in the presence of sodium dodecyl sulfate (SDS), an anionic surfactant, was spectrophotometrically studied. It was found that 0.1-100 mM SDS concentrations stabilize intermediates formed in the peroxidase oxidation of these substrates. The cause of the stabilization is an electrostatic interaction between positively charged intermediates and negatively charged surfactant.
Sigma-Aldrich
o-Dianisidine dihydrochloride, Suitable for use in glucose determination
Sigma-Aldrich
o-Dianisidine dihydrochloride, tablet, 10 mg substrate per tablet
Sigma-Aldrich
o-Dianisidine dihydrochloride, ≥95%
Sigma-Aldrich
o-Dianisidine, peroxidase substrate
Supelco
o-Dianisidine, for spectrophotometric det. of Au, NO2-, Ce(IV), for the detection of Au, Co, Cu, SCN-, V, ≥97.0%