Merck
  • Effect of methylene blue and related redox dyes on monoamine oxidase activity; rat pineal content of N-acetylserotonin, melatonin, and related indoles; and righting reflex in melatonin-primed frogs.
Effect of methylene blue and related redox dyes on monoamine oxidase activity; rat pineal content of N-acetylserotonin, melatonin, and related indoles; and righting reflex in melatonin-primed frogs.
Annals of the New York Academy of Sciences (2007-12-14)
G F Oxenkrug, S O Sablin, P J Requintina
ABSTRACT
The ability of methylene blue (MB) to inhibit the nitric oxide-induced stimulation of N-methyl-D-aspartate receptors has been suggested as a possible mechanism of MB's clinical antidepressant action. This study evaluated the alternative/additional mechanisms of the antidepressant effect of MB on biochemical and behavior levels. Selective inhibition of monoamine oxidase type A (MAO-A) is widely accepted as a major mechanism of the clinical antidepressant effect. MB and the related redox dyes toluidine blue O (TBO), thionine (TN), brilliant cresyl blue (BCB), and toluylene blue (TB) were reversible competitive inhibitors of both MAO-A and MAO-B and were highly selective toward MAO-A. TBO was the most potent inhibitor, followed by TN, BCB, MB, and TB. The dyes studied increased rat pineal N-acetylserotonin (NAS) and melatonin content, in accordance with our previous observations of the stimulating effect of selective inhibition of MAO-A on pineal melatonin biosynthesis. The redox dyes exerted antidepressant-like activity in frogs; that is, they suppressed the righting reflex in melatonin-primed frogs. This study's results indicate that selective inhibition of MAO-A might mediate the clinical antidepressant effect of MB through NAS stimulation and melatonin biosynthesis.
Sigma-Aldrich
N-Acetyl-5-hydroxytryptamine, ≥99% (HPLC), powder