Merck
  • 3-substituted BINOL Schiff bases and their reductive products for catalytic asymmetric addition of diethylzinc to aldehydes.
3-substituted BINOL Schiff bases and their reductive products for catalytic asymmetric addition of diethylzinc to aldehydes.
Chirality (2008-03-14)
Bing Liu, Zhi-Bing Dong, Cao Fang, Hai-Bin Song, Jin-Shan Li
ABSTRACT
Three new chiral 3-substituted BINOL Schiff bases and their reductive 3-arylaminomethyl BINOL products have been synthesized and used for the asymmetric addition reaction of diethylzinc to aldehydes in the presence of titanium tetraisopropoxide. The reaction provided optically active secondary alcohols in high yield (86-100%) and good enantioselectivity (up to 92% ee).
Sigma-Aldrich
Diethylzinc solution, 15 wt. % in toluene
Sigma-Aldrich
Diethylzinc solution, 1.0 M in hexanes
Sigma-Aldrich
Diethylzinc, ≥52 wt. % Zn basis
Sigma-Aldrich
Diethylzinc solution, 1.0 M in heptane