Merck
  • A fluorescence sensor for detection of geranyl pyrophosphate by the chemo-ensemble method.
A fluorescence sensor for detection of geranyl pyrophosphate by the chemo-ensemble method.
The Journal of organic chemistry (2008-12-04)
Kuan-Hung Chen, Jen-Hai Liao, Hsin-Yu Chan, Jim-Min Fang
ABSTRACT
Hexaamide receptor 4 and coumarin phosphate 5 form an ensemble for effective sensing of geranyl pyrophosphate (GPP). Fluorescence resonance energy transfer in the 4 + 5 ensemble diminishes when 5 is replaced by GPP. Receptor 4 binds selectively with GPP over other anions, including fatty acids. The 4-GPP complex in 1:1 stoichiometry is inferred by NMR and fluorescence titrations. Receptor 4 contains a pseudotetrahedral cleft to accommodate GPP via multiple hydrogen bonds, and the two aliphatic chains exert additional hydrophobic interactions.
Sigma-Aldrich
Geranyl pyrophosphate ammonium salt, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥95% (TLC)
Supelco
Geranyl pyrophosphate lithium salt
Sigma-Aldrich
Geranyl pyrophosphate lithium salt, ≥95.0% (TLC)