Merck
  • Effects of manganese exposure on dopamine and prolactin production in rat.
Effects of manganese exposure on dopamine and prolactin production in rat.
Neuroreport (2008-12-06)
Hyeon Yeong Kim, Chae Kwan Lee, Jong Tae Lee, Chan Seok Moon, Sung Chang Ha, Sung Goo Kang, Dae Hwan Kim, Hwi Dong Kim, Jin Hong Ahn, Sung-Bae Lee, Min-Gu Kang
ABSTRACT
Although manganese (Mn) has been shown to increase prolactin (PRL) by decreasing dopamine (DA) in the hypothalamus, the mechanism of Mn-induced regulation of the hypothalamic-hypophyseal-pituitary axis is unclear. We assessed the effects of inhaled Mn on hypothalamic DA and pituitary PRL production and evaluated the role of pituitary-specific transacting factor 1 (Pit-1), a transacting factor of PRL gene, in Mn-induced changes in PRL secretion in the rat brain. Male rats exposed to Mn for 4 or 13 weeks (1.5 mg/m3, 6 h/day, 5 days/week) showed a progressive and significant decrease in hypothalamic DA, whereas PRL and Pit-1 mRNA levels increased in response to Mn exposure. These results suggest that exposure to Mn decreases hypothalamic DA and promotes the production of PRL in the pituitary and that Pit-1 might be a regulator of DA and PRL.
Sigma-Aldrich
Manganese(II) sulfate monohydrate, ACS reagent, ≥98%
Sigma-Aldrich
Manganese sulfate monohydrate, meets USP testing specifications
Sigma-Aldrich
Manganese(II) sulfate monohydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Manganese(II) sulfate monohydrate, BioReagent, suitable for plant cell culture, suitable for cell culture