Merck
  • Palladium-catalyzed cross-coupling reactions of benzyl indium reagents with aryl iodides.
Palladium-catalyzed cross-coupling reactions of benzyl indium reagents with aryl iodides.
The Journal of organic chemistry (2008-12-23)
Louis S Chupak, Joanna P Wolkowski, Yves A Chantigny
ABSTRACT
Herein described is an operationally simple procedure for generating benzyl indium species from readily available benzyl bromides and indium metal followed by in situ palladium-catalyzed coupling with aryl halides. The procedure provides diarylmethanes in modest to excellent yield and tolerates a variety of functional groups in both coupling partners.
Sigma-Aldrich
Diphenylmethane, 99%