Merck
  • Structure elucidation of three diphenyl ether derivatives from the mangrove endophytic fungus SBE-14 from the South China Sea.
Structure elucidation of three diphenyl ether derivatives from the mangrove endophytic fungus SBE-14 from the South China Sea.
Magnetic resonance in chemistry : MRC (2009-02-04)
Fan Liu, Qing Li, Hong Yang, Xiao-Ling Cai, Xue-Kui Xia, Sheng-Ping Chen, Meng-Feng Li, Zhi-Gang She, Yong-Cheng Lin
ABSTRACT
Two new natural products, tenelate A (1) and B (2), together with the known compound, tenellic acid C (3), were isolated from the mangrove endophytic fungus Talaromyces sp. (SBE-14), from the South China Sea. Their structures were elucidated by spectroscopic methods, mainly 1D and 2D NMR techniques.
Sigma-Aldrich
Diphenyl ether, ReagentPlus®, ≥99%
Supelco
Diphenyl ether, Selectophore, ≥99.9%
Sigma-Aldrich
Diphenyl ether, ≥99%, FG
Sigma-Aldrich
Diphenyl ether, ReagentPlus®, 99%